Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022033.4 | NP_071316.1 | 1 | 685 | ||
GenBank mRNA | AB022180.2 | - | 1 | 623 | ||
AK077075.1 | - | 1 | 638 | |||
BC137887.1 | - | 1 | 685 | 8860906 | ||
BC137888.1 | - | 1 | 685 | 8860907 |
Gene Symbol: Oxct2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181859.3 | NP_862907.2 | 1 | 633 | ||
GenBank mRNA | AB049996.1 | - | 1 | 623 | ||
BC113758.1 | - | 1 | 616 | 40047052 |