Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022315.2 | NP_071710.2 | 7 | 804 | ||
GenBank mRNA | AJ249901.1 | - | 7 | 775 | ||
AK030958.1 | - | 7 | 773 | |||
AK159749.1 | - | 7 | 771 | |||
AK160002.1 | - | 7 | 771 | |||
BC019527.1 | - | 7 | 751 | 4191849 |