Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022325.4 | NP_071720.1 | 4 | 620 | ||
GenBank mRNA | AF136277.1 | - | 4 | 630 | ||
AF197479.1 | - | 4 | 547 | |||
AJ242663.1 | - | 4 | 630 | |||
AK002710.1 | - | 4 | 582 | |||
AK004095.1 | - | 4 | 618 | |||
AK008370.1 | - | 4 | 549 | |||
AK010912.1 | - | 4 | 616 | |||
AK150329.1 | - | 4 | 548 | |||
AK150894.1 | - | 4 | 555 | |||
AK150947.1 | - | 4 | 609 | |||
AK151440.1 | - | 4 | 559 | |||
AK151585.1 | - | 4 | 583 | |||
AK151994.1 | - | 4 | 593 | |||
AK153147.1 | - | 4 | 555 | |||
AK159118.1 | - | 4 | 566 | |||
AK159618.1 | - | 4 | 633 | |||
AK159826.1 | - | 4 | 555 | |||
AK160001.1 | - | 4 | 603 | |||
AK160028.1 | - | 4 | 603 | |||
AK160354.1 | - | 4 | 635 | |||
AK166047.1 | - | 4 | 604 | |||
AK170963.1 | - | 4 | 603 | |||
AK184484.1 | - | 2 | 100 | |||
BC008619.1 | - | 4 | 620 | 3588457 |