Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022325.4 | NP_071720.1 | 3, 4 | 591 | ||
GenBank mRNA | AF136277.1 | - | 3, 4 | 601 | ||
AF197479.1 | - | 3, 4 | 518 | |||
AJ242663.1 | - | 3, 4 | 601 | |||
AK002710.1 | - | 3, 4 | 553 | |||
AK004095.1 | - | 3, 4 | 589 | |||
AK008370.1 | - | 3, 4 | 520 | |||
AK010912.1 | - | 3, 4 | 587 | |||
AK150329.1 | - | 3, 4 | 519 | |||
AK150894.1 | - | 3, 4 | 526 | |||
AK150947.1 | - | 3, 4 | 580 | |||
AK151440.1 | - | 3, 4 | 530 | |||
AK151585.1 | - | 3, 4 | 554 | |||
AK151994.1 | - | 3, 4 | 564 | |||
AK153147.1 | - | 3, 4 | 526 | |||
AK159118.1 | - | 3, 4 | 537 | |||
AK159618.1 | - | 3, 4 | 604 | |||
AK159826.1 | - | 3, 4 | 526 | |||
AK160001.1 | - | 3, 4 | 574 | |||
AK160028.1 | - | 3, 4 | 574 | |||
AK160354.1 | - | 3, 4 | 606 | |||
AK166047.1 | - | 3, 4 | 575 | |||
AK170963.1 | - | 3, 4 | 574 | |||
AK184484.1 | - | 1, 2 | 71 | |||
BC008619.1 | - | 3, 4 | 591 | 3588457 |