Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022325.4 | NP_071720.1 | 5 | 766 | ||
GenBank mRNA | AF136277.1 | - | 5 | 776 | ||
AF197479.1 | - | 5 | 693 | |||
AJ242663.1 | - | 5 | 776 | |||
AK002710.1 | - | 5 | 728 | |||
AK004095.1 | - | 5 | 764 | |||
AK008370.1 | - | 5 | 695 | |||
AK010912.1 | - | 5 | 762 | |||
AK150329.1 | - | 5 | 694 | |||
AK150894.1 | - | 5 | 701 | |||
AK150947.1 | - | 5 | 755 | |||
AK151440.1 | - | 5 | 705 | |||
AK151585.1 | - | 5 | 729 | |||
AK151994.1 | - | 5 | 739 | |||
AK153147.1 | - | 5 | 701 | |||
AK159118.1 | - | 5 | 712 | |||
AK159618.1 | - | 5 | 779 | |||
AK159826.1 | - | 5 | 701 | |||
AK160001.1 | - | 5 | 749 | |||
AK160028.1 | - | 5 | 749 | |||
AK160354.1 | - | 5 | 781 | |||
AK166047.1 | - | 5 | 750 | |||
AK170963.1 | - | 5 | 749 | |||
AK181704.1 | - | 1 | 25 | |||
AK185170.1 | - | 1 | 25 | |||
AK185852.1 | - | 1 | 25 | |||
AK186484.1 | - | 2 | 76 | |||
AK192329.1 | - | 1 | 25 | |||
AK215212.1 | - | 1 | 25 | |||
BC008619.1 | - | 5 | 766 | 3588457 |