Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022327.5 | NP_071722.1 | 5 | 783 | ||
GenBank mRNA | AF174296.1 | - | 751 | |||
AK017698.1 | - | 5 | 629 | |||
AK033193.1 | - | 5 | 638 | |||
AK159993.1 | - | 5 | 728 | |||
BC006907.1 | - | 5 | 627 | 3599947 |