Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022327.5 | NP_071722.1 | 4, 5 | 762 | ||
GenBank mRNA | AF174296.1 | - | 730 | |||
AK017698.1 | - | 4, 5 | 608 | |||
AK033193.1 | - | 4, 5 | 617 | |||
AK159993.1 | - | 4, 5 | 707 | |||
BC006907.1 | - | 4, 5 | 606 | 3599947 |