Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252489.1 | NP_001239418.1 | 8 | 1013 | ||
NM_001252490.1 | NP_001239419.1 | 7 | 887 | |||
NM_001252491.1 | NP_001239420.1 | 6 | 1093 | |||
NM_001252492.1 | NP_001239421.1 | 7 | 1256 | |||
NM_001252493.1 | NP_001239422.1 | 7 | 1256 | |||
NM_207530.3 | NP_997413.2 | 21 | 2142 | |||
GenBank mRNA | AB017026.1 | - | 5 | 1137 | ||
AF394063.2 | - | 5 | 448 | |||
AK050641.1 | - | 7 | 811 | |||
AK132676.1 | - | 8 | 1012 | |||
AK133806.1 | - | 5 | 519 | |||
AK168366.1 | - | 7 | 1217 | |||
AK200556.1 | - | 3 | 171 | |||
AY536214.1 | - | 21 | 2021 | |||
BB667982.1 | - | 3 | 215 | |||
BC015075.1 | - | 7 | 977 | |||
BC046466.1 | - | 7 | 1019 | |||
BC057194.1 | - | 6 | 520 | |||
BC076637.1 | - | 7 | 887 | |||
BC092247.1 | - | 6 | 1093 | |||
DV068702.1 | - | 5 | 483 |