Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252489.1 | NP_001239418.1 | 10 | 1353 | ||
NM_001252490.1 | NP_001239419.1 | 9 | 1227 | |||
NM_001252491.1 | NP_001239420.1 | 8 | 1433 | |||
NM_001252492.1 | NP_001239421.1 | 9 | 1596 | |||
NM_001252493.1 | NP_001239422.1 | 9 | 1596 | |||
NM_207530.3 | NP_997413.2 | 23 | 2482 | |||
GenBank mRNA | AB017026.1 | - | 7 | 1477 | ||
AK050641.1 | - | 9 | 1151 | |||
AK132676.1 | - | 10 | 1352 | |||
AK133806.1 | - | 7 | 859 | |||
AK168366.1 | - | 9 | 1557 | |||
AY536214.1 | - | 23 | 2361 | |||
BC015075.1 | - | 9 | 1317 | |||
BC046466.1 | - | 9 | 1359 | |||
BC057194.1 | - | 8 | 860 | |||
BC076637.1 | - | 9 | 1227 | |||
BC092247.1 | - | 8 | 1433 | |||
CO426253.1 | - | 3 | 349 |