Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022419.3 | NP_071864.2 | 2 | 742 | ||
GenBank mRNA | AB030191.1 | - | 2 | 732 | ||
AK003090.1 | - | 2 | 730 | |||
AK037669.1 | - | 2 | 708 | |||
AK080618.1 | - | 2 | 741 | |||
AK155503.1 | - | 2 | 742 | |||
AK205860.1 | - | 1 | 50 | |||
BC026540.1 | - | 2 | 691 | 5050705 |