Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301137.1 | NP_001288066.1 | 2 | 891 | ||
NM_022428.3 | NP_071873.2 | 2 | 891 | |||
GenBank mRNA | AB221658.1 | - | 2 | 144 | ||
AJ271055.1 | - | 2 | 155 | |||
AK035140.1 | - | 2 | 894 | |||
AK053729.1 | - | 2 | 779 | |||
BC130029.1 | - | 2 | 354 | 40140906 |