Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029499.3 | NP_083775.1 | 3 | 305 | ||
GenBank mRNA | AB026045.1 | - | 3 | 292 | ||
AF237914.2 | - | 3 | 279 | |||
AK017928.1 | - | 3 | 285 | |||
AK151045.1 | - | 3 | 298 | |||
AK153495.1 | - | 2 | 149 | |||
AK157137.1 | - | 307 | ||||
AK172650.1 | - | 3 | 277 | |||
BC027496.1 | - | 3 | 261 | 1383416 | ||
CT010170.1 | - | 2 | 173 |