Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033134.3 | NP_149125.1 | 10 | 2477 | ||
GenBank mRNA | AF226683.1 | - | 10 | 2439 | ||
AK047245.1 | - | 8 | 2406 | |||
AK080075.1 | - | 10 | 2459 | |||
AK154097.1 | - | 10 | 2478 | |||
AK158135.1 | - | 10 | 2462 | |||
AK170174.1 | - | 10 | 2547 | |||
AK170786.1 | - | 10 | 2434 | |||
AK170812.1 | - | 9 | 2512 | |||
BC052717.1 | - | 10 | 2446 | 6837339 | ||
BC057566.1 | - | 10 | 2529 | |||
BC080295.1 | - | 10 | 2442 | 30354397 |