Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290437.1 | NP_001277366.1 | 3 | 1240 | ||
NM_033134.3 | NP_149125.1 | 2 | 1524 | |||
GenBank mRNA | AF226683.1 | - | 2 | 1486 | ||
AK047245.1 | - | 2 | 1509 | |||
AK080075.1 | - | 2 | 1506 | |||
AK085439.1 | - | 3 | 1244 | |||
AK154097.1 | - | 2 | 1525 | |||
AK158135.1 | - | 2 | 1509 | |||
AK170174.1 | - | 2 | 1457 | |||
AK170786.1 | - | 2 | 1481 | |||
AK170812.1 | - | 2 | 1495 | |||
BC052717.1 | - | 2 | 1493 | 6837339 | ||
BC057566.1 | - | 2 | 1489 | |||
BC080295.1 | - | 2 | 1489 | 30354397 |