Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133184.2 | NP_573447.2 | 13 | 1715 | ||
GenBank mRNA | AF251267.1 | - | 13 | 1715 | ||
AF498304.1 | - | 2 | 286 |