Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp280b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177475.3 | NP_803426.2 | 2 | 618 | ||
GenBank mRNA | AK031331.1 | - | 2 | 516 | ||
AK144644.1 | - | 2 | 576 | |||
AK144736.1 | - | 2 | 576 | |||
AK157463.1 | - | 1 | 511 | |||
BC094574.1 | - | 2 | 547 | 6856157 | ||
BC117873.1 | - | 3 | 180 | |||
BC117874.2 | - | 2 | 504 | 40095593 |