Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023219.2 | NP_075708.2 | 14 | 1762 | ||
GenBank mRNA | AF251268.1 | - | 14 | 1765 | ||
AF411960.2 | - | 14 | 1682 | |||
AK008310.1 | - | 4 | 462 | |||
AK008368.1 | - | 4 | 462 | |||
AK008500.1 | - | 4 | 461 | |||
AK144296.1 | - | 1705 | ||||
BC109133.1 | - | 14 | 1701 | |||
BC109134.1 | - | 14 | 1701 | |||
BC127612.1 | - | 14 | 1701 | |||
BC127615.1 | - | 14 | 1701 | 40061657 | ||
BC128465.1 | - | 14 | 1701 |