Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022565.2 | NP_072087.2 | 11, 12 | 2944 | ||
GenBank mRNA | AB036838.1 | - | 10, 11 | 2394 | ||
AK132938.1 | - | 6, 7 | 1099 |