Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ireb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022655.3 | NP_073146.2 | 9 | 1209 | ||
GenBank mRNA | BC044665.1 | - | 9 | 1176 | 2647641 | |
CJ093933.1 | - | 5 | 597 | |||
CK636500.1 | - | 5 | 501 |