Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nisch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022656.2 | NP_073147.2 | 3 | 417 | ||
GenBank mRNA | AK004628.1 | - | 3 | 255 | ||
AK014314.1 | - | 3 | 272 | |||
AK036043.1 | - | 3 | 272 | |||
AK049791.1 | - | 3 | 272 | |||
AK054022.1 | - | 2 | 1324 | |||
AK080441.1 | - | 3 | 294 | |||
AK086880.1 | - | 3 | 272 | |||
AK088377.1 | - | 2 | 1304 | |||
AK122412.1 | - | 3 | 392 | |||
AK160990.1 | - | 3 | 291 | |||
BC003270.1 | - | 3 | 279 | |||
BC066838.1 | - | 3 | 275 | |||
CJ106892.1 | - | 3 | 418 |