Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nisch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022656.2 | NP_073147.2 | 21 | 4425 | ||
GenBank mRNA | AF144133.1 | - | 2 | 106 | ||
AF315344.1 | - | 17 | 3878 | |||
AK122412.1 | - | 19 | 3923 | |||
AY032852.1 | - | 4 | 560 | |||
BC089329.1 | - | 16 | 3644 | |||
BC108364.1 | - | 16 | 3620 | |||
BC144980.1 | - | 17 | 3754 |