Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026080.2 | NP_080356.1 | 4 | 326 | ||
GenBank mRNA | AK003117.1 | - | 4 | 310 | ||
AK007338.1 | - | 4 | 310 | |||
AK010590.1 | - | 4 | 307 | |||
AK012461.1 | - | 4 | 315 | |||
AK014099.1 | - | 4 | 313 | |||
BC029262.1 | - | 4 | 300 | 4911699 | ||
BY100364.1 | - | 4 | 329 |