Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270428.1 | NP_001257357.1 | 2 | 278 | ||
NM_002315.2 | NP_002306.1 | 2 | 610 | |||
NR_073006.1 | 2 | 610 | ||||
XM_006718228.3 | XP_006718291.1 | 2 | 1656 | |||
XM_011520098.2 | XP_011518400.1 | 2 | 145 | |||
XM_011520099.2 | XP_011518401.1 | 2 | 162 | |||
GenBank mRNA | BC039512.1 | - | 2 | 278 | ||
BC069673.1 | - | 2 | 152 | 7262648 | ||
BC069752.1 | - | 2 | 486 | 7262660 | ||
BC069793.1 | - | 2 | 486 | 7262577 | ||
BC096056.3 | - | 2 | 120 | 40000075 | ||
BC096057.3 | - | 2 | 119 | 40000076 | ||
BC096058.1 | - | 2 | 120 | 40000078 | ||
M26682.1 | - | 2 | 591 | |||
X64244.1 | - | 2 | 94 | |||
X64245.1 | - | 1 | 108 |