Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271472.1 | NP_001258401.1 | 19 | 2541 | ||
NM_001271473.1 | NP_001258402.1 | 16 | 2208 | |||
NM_022723.3 | NP_073560.2 | 18 | 2451 | |||
GenBank mRNA | AF276425.1 | - | 18 | 2318 | ||
AK041590.1 | - | 18 | 2434 | |||
AK083722.1 | - | 2 | 400 | |||
BC066066.1 | - | 19 | 2499 | 6850999 | ||
BC080278.1 | - | 16 | 2176 | 30354974 |