Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199118.1 | NP_001186047.1 | 15 | 2068 | ||
NM_022883.3 | NP_075021.1 | 15 | 2145 | |||
GenBank mRNA | AF286724.1 | - | 15 | 2082 | ||
AK033647.1 | - | 15 | 2096 | |||
AK049363.1 | - | 15 | 2061 | |||
AK163833.1 | - | 15 | 2060 | |||
AK220228.1 | - | 15 | 2145 | |||
BC026729.1 | - | 15 | 2052 | |||
BC117882.1 | - | 14 | 2009 | 40096038 | ||
BC117883.1 | - | 14 | 2009 | 40096041 |