Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289575.1 | NP_001276504.1 | 14 | 1636 | ||
NM_001289576.1 | NP_001276505.1 | 14 | 1633 | |||
NM_022887.4 | NP_075025.2 | 14 | 1604 | |||
GenBank mRNA | AB047561.1 | - | 12 | 1403 | ||
AJ271912.1 | - | 14 | 1628 | |||
AK030318.1 | - | 14 | 1600 | |||
AK147428.1 | - | 14 | 1624 | |||
BC052399.1 | - | 13 | 1557 | 5720967 |