Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022985.6 | NP_075361.2 | 3 | 354 | ||
GenBank mRNA | AJ251508.1 | - | 260 | |||
AK002775.1 | - | 3 | 355 | |||
AK009403.1 | - | 2 | 212 | |||
AK144902.1 | - | 2 | 204 | |||
AK212602.1 | - | 1 | 81 | |||
AK216328.1 | - | 1 | 81 | |||
BC010683.1 | - | 2 | 215 | 2647905 |