Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022994.3 | NP_075370.2 | 11 | 1354 | ||
GenBank mRNA | AJ250375.1 | - | 11 | 1098 | ||
AK014889.1 | - | 11 | 1024 | |||
AK032275.1 | - | 11 | 1026 | |||
BC019566.1 | - | 11 | 1354 | 4235517 |