Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164533.1 | NP_001158005.1 | 11, 12 | 1428 | ||
NM_022994.3 | NP_075370.2 | 12, 13 | 1518 | |||
GenBank mRNA | AJ250375.1 | - | 12, 13 | 1262 | ||
AK014889.1 | - | 12, 13 | 1188 | |||
AK021250.1 | - | 7, 8 | 724 | |||
AK032275.1 | - | 12, 13 | 1190 | |||
AK088960.1 | - | 11, 12 | 1098 | |||
BC019566.1 | - | 12, 13 | 1518 | 4235517 |