Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LMO7 SIRNA Assays ›
Gene Symbol:
LMO7
Gene Name:
LIM domain 7
Gene Aliases:
FBX20, FBXO20, LOMP
Chromosome Location:
Chr.13: 75620434 - 75859870 on Build GRCh38
UniGene ID:
Hs.207631
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8238
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LMO7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306080.1
NP_001293009.1
8
1096
NM_005358.5
NP_005349.3
9
2142
NM_015842.2
NP_056667.2
5
787
XM_011535075.2
XP_011533377.1
7
657
XM_011535078.1
XP_011533380.1
7
657
XM_011535080.2
XP_011533382.1
7
657
XM_011535082.1
XP_011533384.1
7
657
XM_011535086.1
XP_011533388.1
7
657
XM_011535094.1
XP_011533396.1
5
516
XM_011535095.1
XP_011533397.1
7
657
XM_017020574.1
XP_016876063.1
7
657
XM_017020575.1
XP_016876064.1
7
657
XM_017020576.1
XP_016876065.1
7
657
XM_017020577.1
XP_016876066.1
7
657
XM_017020578.1
XP_016876067.1
7
657
XM_017020579.1
XP_016876068.1
7
657
XM_017020580.1
XP_016876069.1
7
657
XM_017020581.1
XP_016876070.1
6
579
XM_017020582.1
XP_016876071.1
7
657
XM_017020583.1
XP_016876072.1
7
657
XM_017020584.1
XP_016876073.1
6
579
XM_017020585.1
XP_016876074.1
7
657
XM_017020586.1
XP_016876075.1
7
657
XM_017020587.1
XP_016876076.1
7
657
XM_017020588.1
XP_016876077.1
7
657
XM_017020589.1
XP_016876078.1
6
579
XM_017020590.1
XP_016876079.1
7
657
XM_017020591.1
XP_016876080.1
6
740
XM_017020592.1
XP_016876081.1
6
623
XM_017020593.1
XP_016876082.1
6
740
XM_017020594.1
XP_016876083.1
6
623
XM_017020595.1
XP_016876084.1
6
740
XM_017020596.1
XP_016876085.1
7
657
XM_017020597.1
XP_016876086.1
7
657
XM_017020598.1
XP_016876087.1
6
740
XM_017020599.1
XP_016876088.1
6
740
XM_017020600.1
XP_016876089.1
7
657
XM_017020601.1
XP_016876090.1
6
740
XM_017020602.1
XP_016876091.1
6
623
XM_017020603.1
XP_016876092.1
6
740
XM_017020604.1
XP_016876093.1
6
740
GenBank mRNA
AB020665.2
-
5
717
AF330045.1
-
9
2141
AK055465.1
-
1
34
AK092052.1
-
8
1061
AK304051.1
-
6
629
DB064574.1
-
1
117
FJ711162.1
-
8
726
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of signaling
regulation of cell adhesion
ubiquitin-protein transferase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline