Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023047.2 | NP_075534.1 | 2 | 331 | ||
GenBank mRNA | AF249295.1 | - | 2 | 495 | ||
AK044866.1 | - | 2 | 203 | |||
BC065054.1 | - | 2 | 331 | 5709296 | ||
BC103789.1 | - | 2 | 158 |