Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174069.1 | NP_001167540.1 | 9 | 1579 | ||
NM_177398.3 | NP_796372.1 | 9 | 1768 | |||
XM_011509538.2 | XP_011507840.1 | 7 | 1236 | |||
GenBank mRNA | AK122800.1 | - | 9 | 1579 | ||
AK127724.1 | - | 9 | 1768 | |||
BC066353.1 | - | 7 | 1019 |