Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023906.3 | NP_076395.2 | 3, 4 | 534 | ||
GenBank mRNA | AF155354.1 | - | 2, 3 | 349 | ||
AK010315.1 | - | 3, 4 | 470 | |||
AK028489.1 | - | 4, 5 | 640 | |||
AK038086.1 | - | 3, 4 | 515 | |||
AK143580.1 | - | 3, 4 | 469 | |||
BC023086.1 | - | 3, 4 | 468 | 4947610 |