Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290989.1 | NP_001277918.1 | 8 | 1435 | ||
NM_023051.5 | NP_075538.1 | 9 | 1465 | |||
GenBank mRNA | AJ289016.1 | - | 9 | 1296 | ||
AK173059.1 | - | 8 | 1475 | |||
BC029027.1 | - | 4 | 550 | |||
BC053843.1 | - | 8 | 1404 | 6315355 | ||
CO433558.1 | - | 3 | 438 |