Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023055.2 | NP_075542.2 | 7 | 1505 | ||
NM_023449.3 | NP_075938.2 | 6 | 1075 | |||
GenBank mRNA | AB026489.1 | - | 7 | 1478 | ||
AK002557.1 | - | 6 | 1076 | |||
AK004710.1 | - | 3 | 748 | |||
AK039491.1 | - | 7 | 1493 | |||
AK075800.1 | - | 7 | 1508 | |||
AK075813.1 | - | 7 | 1506 | |||
AK084801.1 | - | 7 | 1506 | |||
AK169007.1 | - | 7 | 1496 | |||
AK170057.1 | - | 7 | 1491 | |||
BC070458.1 | - | 6 | 1426 | 30603578 |