Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 5 | 910 | ||
NM_001164208.1 | NP_001157680.1 | 5 | 558 | |||
NM_001164209.2 | NP_001157681.1 | 6 | 766 | |||
NM_001286651.1 | NP_001273580.1 | 6 | 763 | |||
NM_001286652.1 | NP_001273581.1 | 5 | 670 | |||
NM_023056.4 | NP_075543.1 | 5 | 1159 | |||
GenBank mRNA | AB031386.1 | - | 5 | 719 | ||
AF115426.1 | - | 5 | 631 | |||
AK007408.1 | - | 5 | 565 | |||
AK156042.1 | - | 5 | 911 | |||
BC019539.1 | - | 4 | 528 | 4237639 | ||
BC022166.1 | - | 5 | 1174 | 3582123 | ||
BQ571258.1 | - | 5 | 550 | |||
CT010176.1 | - | 4 | 523 | |||
DV047496.1 | - | 5 | 597 | |||
DV059241.1 | - | 5 | 634 |