Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164791.1 | NP_001158263.1 | 8 | 787 | ||
NM_023057.5 | NP_075544.1 | 8 | 851 | |||
NM_178084.4 | NP_835185.2 | 8 | 787 | |||
GenBank mRNA | AB049731.1 | - | 7 | 634 | ||
AB049732.1 | - | 7 | 634 | |||
AK045444.1 | - | 8 | 712 | |||
AK087363.1 | - | 8 | 727 | |||
AK132186.1 | - | 8 | 852 | |||
AK162819.1 | - | 8 | 652 | |||
BC023718.1 | - | 8 | 732 | 5346593 |