Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023060.3 | NP_075547.1 | 7 | 1665 | ||
GenBank mRNA | AF268871.1 | - | 7 | 1629 | ||
AF283518.1 | - | 7 | 1620 | |||
AK028424.1 | - | 7 | 1647 | |||
AK087501.1 | - | 8 | 1733 | |||
AK147924.1 | - | 7 | 1634 | |||
AK171885.1 | - | 7 | 1645 | |||
AK212932.1 | - | 2 | 174 | |||
BC024915.1 | - | 7 | 1639 | 3158026 | ||
BC099675.1 | - | 7 | 1623 | 5011896 |