Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cubn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081084.2 | NP_001074553.1 | 53 | 8285 | ||
GenBank mRNA | BC026593.1 | - | 2 | 85 | ||
CK391475.1 | - | 2 | 85 |