Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017407.1 | NP_001017407.1 | 2 | 296 | ||
NM_001017409.1 | NP_001017409.1 | 2 | 296 | |||
NM_001017419.2 | NP_001017419.1 | 3 | 378 | |||
NM_001017433.2 | NP_001017433.1 | 3 | 378 | |||
NM_001017441.2 | NP_001017441.2 | 3 | 378 | |||
NM_023064.3 | NP_075551.3 | 2 | 296 | |||
GenBank mRNA | AF257502.1 | - | 2 | 296 | ||
AF257503.1 | - | 2 | 296 | |||
AF521591.1 | - | 2 | 309 | |||
AF521592.1 | - | 2 | 309 | |||
AJ249392.1 | - | 1 | 166 | |||
AK038284.1 | - | 1 | 166 | |||
AK088524.1 | - | 1 | 166 | |||
AK132793.1 | - | 3 | 379 | |||
AY243457.1 | - | 2 | 296 | |||
AY243458.1 | - | 1 | 144 | |||
AY243459.1 | - | 1 | 166 | |||
BU937744.1 | - | 3 | 391 | |||
EU622858.1 | - | 2 | 213 |