Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023146.2 | NP_075635.1 | 13, 14 | 1707 | ||
GenBank mRNA | AJ271458.1 | - | 13, 14 | 1605 | ||
AK016583.1 | - | 13, 14 | 1607 | |||
AK035897.1 | - | 13, 14 | 1604 | |||
AK050150.1 | - | 9, 10 | 1158 | |||
AK163271.1 | - | 1412 | ||||
BC033503.1 | - | 13, 14 | 1601 |