Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem254a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025311.3 | NP_079587.1 | 279 | |||
NM_026679.2 | NP_080955.1 | 448 | ||||
GenBank mRNA | AK002342.1 | - | 2 | 267 | ||
AK002441.1 | - | 2 | 379 | |||
AK008299.1 | - | 2 | 239 | |||
BC031854.1 | - | 2 | 332 | 4194400 | ||
BY270140.1 | - | 280 |
Gene Symbol: Tmem254b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270495.1 | NP_001257424.1 | 2 | 432 | ||
NM_001270496.1 | NP_001257425.1 | 2 | 190 | |||
GenBank mRNA | BY713360.1 | - | 2 | 432 | ||
CK793021.1 | - | 2 | 188 |
Gene Symbol: Tmem254c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270498.1 | NP_001257427.1 | 448 | |||
NM_001270499.1 | NP_001257428.1 | 268 | ||||
GenBank mRNA | CK128852.1 | - | 363 | |||
CX201482.1 | - | 268 |