Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025316.2 | NP_079592.2 | 2, 3 | 230 | ||
GenBank mRNA | AK002279.1 | - | 2, 3 | 232 | ||
AK004372.1 | - | 2, 3 | 230 | |||
AK005521.1 | - | 2, 3 | 232 | |||
AK013223.1 | - | 2, 3 | 232 | |||
AK018562.1 | - | 2, 3 | 237 | |||
AK088061.1 | - | 2, 3 | 239 | |||
BC025155.1 | - | 2, 3 | 223 | 4024605 |