Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168470.1 | NP_001161942.1 | 2 | 437 | ||
NM_025318.3 | NP_079594.1 | 2 | 388 | |||
GenBank mRNA | AK002385.1 | - | 2 | 373 | ||
AK002668.1 | - | 364 | ||||
AK003198.1 | - | 2 | 383 | |||
AK004149.1 | - | 2 | 384 | |||
AK012769.1 | - | 2 | 373 | |||
BC022104.1 | - | 2 | 359 | 5101119 | ||
DV661517.1 | - | 2 | 372 |