Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130142.1 | NP_001123614.1 | 9 | 1165 | ||
NM_014622.4 | NP_055437.2 | 8 | 1050 | |||
NM_198315.2 | NP_938057.1 | 8 | 1050 | |||
XM_011542828.2 | XP_011541130.1 | 9 | 1276 | |||
GenBank mRNA | AF002672.1 | - | 7 | 962 | ||
AK291698.1 | - | 8 | 1004 | |||
AK295247.1 | - | 9 | 1244 | |||
AK314934.1 | - | 1 | 15 | |||
AY366503.1 | - | 8 | 1050 | |||
AY366505.1 | - | 8 | 1050 | |||
AY366506.1 | - | 9 | 1165 | |||
AY366508.1 | - | 9 | 1165 | |||
BC001234.1 | - | 8 | 1027 | 3461486 | ||
BT006655.1 | - | 7 | 956 |