Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289531.1 | NP_001276460.1 | 8 | 1219 | ||
NM_023149.3 | NP_075638.2 | 9 | 1187 | |||
GenBank mRNA | AB046738.1 | - | 9 | 1128 | ||
AK003779.1 | - | 9 | 1108 | |||
AK131124.1 | - | 8 | 1099 | |||
AK149751.1 | - | 8 | 1143 | |||
AK151013.1 | - | 9 | 1109 | |||
AK152442.1 | - | 8 | 1184 | |||
AK152729.1 | - | 8 | 1157 | |||
AK153040.1 | - | 8 | 1158 | |||
AK159129.1 | - | 8 | 1186 | |||
AK159739.1 | - | 9 | 1114 | |||
AK168029.1 | - | 9 | 1126 | |||
AK168652.1 | - | 9 | 1138 | |||
AK168856.1 | - | 7 | 876 | |||
AK172034.1 | - | 9 | 1109 | |||
BC005532.1 | - | 9 | 1118 | 3496319 |