Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025323.2 | NP_079599.1 | 2 | 317 | ||
GenBank mRNA | AK002375.1 | - | 2 | 365 | ||
AK003683.1 | - | 2 | 373 | |||
AK034275.1 | - | 2 | 365 | |||
AK151648.1 | - | 2 | 360 | |||
BC019535.1 | - | 2 | 317 | 4458983 |