Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130142.1 | NP_001123614.1 | 10 | 1246 | ||
NM_014622.4 | NP_055437.2 | 9 | 1131 | |||
NM_198315.2 | NP_938057.1 | 9 | 1131 | |||
XM_011542828.2 | XP_011541130.1 | 10 | 1357 | |||
GenBank mRNA | AF002672.1 | - | 8 | 1043 | ||
AK291698.1 | - | 9 | 1085 | |||
AK295247.1 | - | 10 | 1325 | |||
AK314934.1 | - | 2 | 96 | |||
AY366503.1 | - | 9 | 1131 | |||
AY366505.1 | - | 9 | 1131 | |||
AY366506.1 | - | 10 | 1246 | |||
AY366508.1 | - | 10 | 1246 | |||
BC001234.1 | - | 9 | 1108 | 3461486 | ||
BT006655.1 | - | 8 | 1037 |