Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cystm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081365.1 | NP_001074834.1 | 2 | 374 | ||
GenBank mRNA | AK002512.1 | - | 247 | |||
AK003470.1 | - | 75 | ||||
AK028217.1 | - | 245 | ||||
AK089711.1 | - | 2 | 236 | |||
AK167679.1 | - | 279 | ||||
BC028765.1 | - | 301 | 1195860 |