Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctex1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025329.3 | NP_079605.1 | 2 | 251 | ||
GenBank mRNA | AK002589.1 | - | 2 | 210 | ||
AK004191.1 | - | 2 | 179 | |||
AK007178.1 | - | 2 | 239 | |||
AK010027.1 | - | 2 | 209 | |||
AK010464.1 | - | 2 | 207 | |||
BC059716.1 | - | 2 | 252 | 6774264 |